EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
The effect of allergen-induced bronchoconstriction on concentration of 5-oxo-ETE in exhaled breath condensate of house dust mite-allergic patients.
PMID 28763131
Abstract
Arachidonic acid metabolites regulate several aspects of airway function including inflammation, muscle contraction and mucous secretion. The aim of this study was to evaluate concentration of selected 5-lipoxygenase- and cyclooxygenase-derived eicosanoids in exhaled breath condensate (EBC) during allergen-induced bronchoconstriction. The study was performed on 24 allergic rhinitis/asthma patients sensitized to a house dust mite (HDM) Dermatophagoides pteronyssinus (Dp) and 13 healthy controls (HCs). Bronchial challenge with Dp extract was performed only in the allergic patients. EBC samples were collected before (T Significant bronchoconstriction after Dp challenge was demonstrated in 15 patients (Rs), while in 9 patients (NRs) no asthmatic response could be detected. At T Our study demonstrates significant up-regulation of 5-oxo-ETE synthesis in HDM-allergic patients and indicates possible involvement of that mediator in the pathogenesis of allergic asthma.