EMAIL THIS PAGE TO A FRIEND
Food chemistry
Determination of steroid hormones in fish tissues by microwave-assisted extraction coupled to ultra-high performance liquid chromatography tandem mass spectrometry.
PMID 28763944
Abstract
Steroid hormones produce adverse effects on biota as well as bioaccumulation in fish and seafood, making it necessary to develop methodologies to evaluate these compounds in samples related to the food chain. This work presents an analytical method for evaluating 15 steroid hormones in fish tissue. It is based on microwave-assisted extraction and solid-phase extraction coupled to ultra-high-performance liquid chromatography tandem mass spectrometry (MAE-SPE-UHPLC-MS/MS). The proposed method shows appropriate detection limits (0.14-49.0ngg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8115
Coronatine, from Pseudomonas syringae pv Glycinea, >95% (HPLC)
C18H25NO4