EMAIL THIS PAGE TO A FRIEND
Food chemistry
A novel magnetic ion imprinted polymer as a selective magnetic solid phase for separation of trace lead(II) ions from agricultural products, and optimization using a Box-Behnken design.
PMID 28763996
Abstract
In this work, a magnetic ion-imprinted polymer (Fe
Related Materials