EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Synthesis of disaccharides using β-glucosidases from Aspergillus niger, A. awamori and Prunus dulcis.
PMID 28766088
Abstract
Glucose conversion into disaccharides was performed with β-glucosidases from Prunus dulcis (β-Pd), Aspergillus niger (β-An) and A. awamori (β-Aa), in reactions containing initial glucose of 700 and 900 g l The reactions' time courses were followed regarding glucose and product concentrations. In all cases, there was a predominant formation of gentiobiose over cellobiose and also of oligosaccharides with a higher molecular mass. For reactions containing 700 g glucose l β-Pd and the fungal β-An and β-Aa β-glucosidases present quite dissimilar kinetics and selective properties regarding the synthesis of disaccharides; while β-Pd showed the highest productivity for gentiobiose synthesis, β-An presented the highest specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3000
β-Gentiobiose, ≥85% (remainder primarily α-anomer)
C12H22O11