EMAIL THIS PAGE TO A FRIEND
Faraday discussions
Supramolecular chiral self-assemblies of Gly-Pro dipeptides on metallic fcc(110) surfaces.
PMID 28766638
Abstract
Adsorption of the Glycine-Proline (Gly-Pro) dipeptide has been investigated using surface science complementary techniques on Au(110) and Ag(110), showing some interesting differences both in the chemical form and surface organization of the adsorbed peptide. On Au(110), Gly-Pro mainly adsorbs in neutral form (COOH/NH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3002
Gly-Pro
C7H12N2O3