EMAIL THIS PAGE TO A FRIEND
Immunology
Programmed death 1 is highly expressed on CD8
PMID 28767147
Abstract
Growing evidence points to a deregulated response to Epstein-Barr virus (EBV) in the central nervous system of patients with multiple sclerosis (MS) as a possible cause of disease. We have investigated the response of a subpopulation of effector CD8