EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Light-Enhanced Antibacterial Activity of Graphene Oxide, Mainly via Accelerated Electron Transfer.
PMID 28771330
Abstract
Before graphene derivatives can be exploited as next-generation antimicrobials, we must understand their behavior under environmental conditions. Here, we demonstrate how exposure to simulated sunlight significantly enhances the antibacterial activity of graphene oxide (GO) and reveal the underlying mechanism. Our measurements of reactive oxygen species (ROS) showed that only singlet oxygen (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
655546
1,6-Bis(p-carboxyphenoxy)hexane, 90%
C20H22O6