EMAIL THIS PAGE TO A FRIEND
Scientific reports
ADAMTS-13 regulates neutrophil recruitment in a mouse model of invasive pulmonary aspergillosis.
PMID 28775254
Abstract
Von Willebrand factor (VWF) is secreted as an acute phase protein during inflammation. ADAMTS-13 regulates the size and prothrombotic activity of VWF by it's specific proteolytic activity. To determine the relevance of this regulatory pathway for the innate inflammatory response by polymorphonuclear neutrophils (PMN), we employed a mouse model of invasive pulmonary aspergillosis (IPA) where PMN functionality is crucial for fungal clearance and survival. IPA was induced by intratracheal application of Aspergillus fumigatus (A. fumigatus) conidia in wildtype (129/Sv/Pas) or ADAMTS-13 deficient (Adamts13