EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Autonomous and Continuous Stimuli-Responsive Polymer Surface for Antibacterial Application through Enzymatic Self-Propagating Reactions.
PMID 28776785
Abstract
Stimuli-responsive polymer materials inspired by biological materials have invoked increasing research interest; however, they still suffer from limitations such as finite amplified responses and poor sensitivity of the unstimulated parts. Herein, a new strategy for creating H