EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Flower-like MoSe
PMID 28777498
Abstract
Sodium-ion batteries (SIBs) have caught considerable attention in last few years owing to the abundance of sodium in comparison to lithium. The commercial graphite anode is demonstrated unsuitable as an anode material for SIBs due to the larger radius of Na