EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Controlled Isotropic and Anisotropic Shell Growth in β-NaLnF
PMID 28777550
Abstract
Precise morphology and composition control is vital for designing multifunctional lanthanide-doped core/shell nanocrystals. Herein, we report controlled isotropic and anisotropic shell growth techniques in hexagonal sodium rare-earth tetrafluoride (β-NaLnF