EMAIL THIS PAGE TO A FRIEND
Physics in medicine and biology
Characterization of breast calcification types using dual energy x-ray method.
PMID 28777746
Abstract
Calcifications are products of mineralization whose presence is usually associated with pathological conditions. The minerals mostly seen in several diseases are calcium oxalate (CaC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
455997
Calcium oxalate, ≥99.9% trace metals basis
C2CaO4