EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Histone H4K20 tri-methylation at late-firing origins ensures timely heterochromatin replication.
PMID 28778956
Abstract
Among other targets, the protein lysine methyltransferase PR-Set7 induces histone H4 lysine 20 monomethylation (H4K20me1), which is the substrate for further methylation by the Suv4-20h methyltransferase. Although these enzymes have been implicated in control of replication origins, the specific contribution of H4K20 methylation to DNA replication remains unclear. Here, we show that H4K20 mutation in mammalian cells, unlike in
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU130861 MISSION® esiRNA, esiRNA human LRWD1 (esiRNA1)