EMAIL THIS PAGE TO A FRIEND
Water research
Counter-diffusion biofilms have lower N
PMID 28780360
Abstract
The goal of this study was to investigate the effectiveness of a membrane-aerated biofilm reactor (MABR), a representative of counter-current substrate diffusion geometry, in mitigating nitrous oxide (N