EMAIL THIS PAGE TO A FRIEND
Oncology letters
MicroRNA-150 functions as an antioncogenic regulator in osteosarcoma.
PMID 28781686
Abstract
Numerous studies have demonstrated that microRNAs (miRs) are involved in several physiological and pathological processes, and participate in cancer initiation and progression. The abnormal expression of miR-150 has been reported in numerous types of human cancer. However, at present there are no studies of miR-150 in osteosarcoma (OS). Reverse transcription-quantitative polymerase chain reaction was performed to measure miR-150 expression levels in OS tissues and cell lines. Subsequent to transfection with miR-150 mimics or zinc finger E-box binding homeobox 1 (ZEB1) small interfering RNA, an MTT assay, Transwell migration and invasion assays, western blotting and a Dual-Luciferase reporter assay were performed in human OS cell lines. The present study revealed that miR-150 was downregulated in OS tissues and cell lines. In addition, the expression levels of miR-150 were correlated with the clinical stage and degree of distant metastasis of patients with OS. In addition, ZEB1 was identified as a direct target of miR-150
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU151901 MISSION® esiRNA, esiRNA human ZEB1 (esiRNA1)