EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Anti-inflammatory effect of Cortex Eucommiae via modulation of the toll-like receptor 4 pathway in lipopolysaccharide-stimulated RAW 264.7 macrophages.
PMID 28782620
Abstract
Cortex Eucommiae (CE), the bark of Eucommia ulmoides Oliv., has been traditionally used for its kidney-tonifying and bone- and tendon-enhancing properties in Korea, China, and Japan. CE has been historically prescribed for inflammatory conditions such as arthritis of the knee and ankle. Although CE has recently been shown to suppress inflammation in scientific studies, whether this effect involves modulation of the toll-like receptor 4 (TLR-4) pathway is currently unknown. The modulatory effect of CE on the TLR-4 pathway, both myeloid differentiation primary response gene 88 (Myd88)-dependent and independent, was investigated through real-time reverse transcriptase-polymerase chain reaction (RT-PCR), western blotting, and a reporter gene assay in lipopolysaccharide (LPS)-stimulated RAW 264.7 murine macrophages. CE dose-dependently inhibited nitric oxide production without significant cytotoxicity with an IC CE down-regulated both Myd88-dependent and independent TLR-4 pathways, thus exerting anti-inflammatory effects. These results suggest that CE may be used as a potential therapeutic agent against chronic inflammatory diseases.