EMAIL THIS PAGE TO A FRIEND
Magnetic resonance imaging
Transverse relaxation of cerebrospinal fluid depends on glucose concentration.
PMID 28782676
Abstract
To evaluate the biophysical processes that generate specific T CSF T Ex vivo CSF T This study opens the possibility of monitoring glucose regulation of CSF at the resolution of MRI by quantitating T