EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Epiregulin and EGFR interactions are involved in pain processing.
PMID 28783046
Abstract
The EGFR belongs to the well-studied ErbB family of receptor tyrosine kinases. EGFR is activated by numerous endogenous ligands that promote cellular growth, proliferation, and tissue regeneration. In the present study, we have demonstrated a role for EGFR and its natural ligand, epiregulin (EREG), in pain processing. We show that inhibition of EGFR with clinically available compounds strongly reduced nocifensive behavior in mouse models of inflammatory and chronic pain. EREG-mediated activation of EGFR enhanced nociception through a mechanism involving the PI3K/AKT/mTOR pathway and matrix metalloproteinase-9. Moreover, EREG application potentiated capsaicin-induced calcium influx in a subset of sensory neurons. Both the EGFR and EREG genes displayed a genetic association with the development of chronic pain in several clinical cohorts of temporomandibular disorder. Thus, EGFR and EREG may be suitable therapeutic targets for persistent pain conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2731
AMG 9810, ≥98% (HPLC), powder
C21H23NO3
H4415
HC-030031, ≥98% (HPLC), powder
C18H21N5O3
P215
PD 98,059, solid
C16H13NO3