EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Development of resistance to photodynamic therapy (PDT) in human breast cancer cells is photosensitizer-dependent: Possible mechanisms and approaches for overcoming PDT-resistance.
PMID 28784290
Abstract
Here we report on the induction of resistance to photodynamic therapy (PDT) in the ABCG2-high human breast cancer cell line MA11 after repetitive PDT, using either Pheophorbide A (PhA) or di-sulphonated meso-tetraphenylchlorin (TPCS
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160415
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine, ≥99%
C20H14N4