EMAIL THIS PAGE TO A FRIEND
Cancer science
Contrasting effects of cyclophosphamide on anti-CTL-associated protein 4 blockade therapy in two mouse tumor models.
PMID 28787548
Abstract
Immune checkpoint blockade is a promising anticancer therapy, but must be used in combination with other anticancer therapies to increase its therapeutic efficacy. Cyclophosphamide (CP) is a chemotherapeutic drug that shows immune-modulating effects. In this study, we examined the effect of CP on anti-CTL-associated protein 4 (CTLA-4) blockade therapy in two mouse tumor models. Drastic tumor regression was observed in the CT26 colon carcinoma model after i.p. injection of CP (100 mg/kg) followed by anti-CTLA-4 antibody. However, administration in the reverse order increased apoptosis in tumor-specific CD8