EMAIL THIS PAGE TO A FRIEND
Chemosphere
Estrogenic activity, chemical levels and health risk assessment of municipal distribution point water from Pretoria and Cape Town, South Africa.
PMID 28787686
Abstract
Endocrine disrupting chemicals (EDCs) are ubiquitous in the environment and have been detected in drinking water from various countries. Although various water treatment processes can remove EDCs, chemicals can also migrate from pipes that transport water and contaminate drinking water. This study investigated the estrogenic activity in drinking water from various distribution points in Pretoria (City of Tshwane) (n = 40) and Cape Town (n = 40), South Africa, using the recombinant yeast estrogen screen (YES) and the T47D-KBluc reporter gene assay. The samples were collected seasonally over four sampling periods. The samples were also analysed for bisphenol A (BPA), nonylphenol (NP), di(2-ethylhexyl) adipate (DEHA), dibutyl phthalate (DBP), di(2-ethylhexyl) phthalate (DEHP), diisononylphthalate (DINP), 17β-estradiol (E