EMAIL THIS PAGE TO A FRIEND
Oncology letters
Treatment with a JNK inhibitor increases, whereas treatment with a p38 inhibitor decreases, H
PMID 28789403
Abstract
Oxidative stress induces apoptosis in endothelial cells (ECs). Reactive oxygen species (ROS) promote cell death by regulating the activity of various mitogen-activated protein kinases (MAPKs) in ECs. The present study investigated the effects of MAPK inhibitors on cell survival and glutathione (GSH) levels upon H