EMAIL THIS PAGE TO A FRIEND
Oncology letters
p53 performs an essential role in mediating the oncogenic stimulus triggered by loss of expression of neurofibromatosis type 2 during
PMID 28789444
Abstract
The loss of the tumor suppressor