EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A hybrid molecule of a GFP chromophore analogue and cholestene as a viscosity-dependent and cholesterol-responsive fluorescent sensor.
PMID 28792046
Abstract
Diarylmethylenated and cholestene (or -tane)-hybrid analogues of the GFP chromophore, namely, Ch-DAINs were successfully synthesised by a condensation reaction between methyl imidates and N-(diarylmethylene)glycinates. Among the Ch-DAINs synthesised, a diphenyl-type analogue showed viscosity-dependent and cholesterol-responsive fluorescent properties. It showed a nearly linear increase of the fluorescence emission in triglycerides and vesicles as the amount of cholesterol was increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1978000
Tricaprylin, European Pharmacopoeia (EP) Reference Standard
C27H50O6