EMAIL THIS PAGE TO A FRIEND
Clinical laboratory
Performance of Clinical Features, Acute Phase Reactants and Group A Streptococcus Rapid Test in Evaluation of the Etiologic Agents for Tonsillopharyngitis in Children.
PMID 28792720