EMAIL THIS PAGE TO A FRIEND
Biophysical journal
Critical Influence of Cosolutes and Surfaces on the Assembly of Serpin-Derived Amyloid Fibrils.
PMID 28793213
Abstract
Many proteins and peptides self-associate into highly ordered and structurally similar amyloid cross-β aggregates. This fibrillation is critically dependent on properties of the protein and the surrounding environment that alter kinetic and thermodynamic equilibria. Here, we report on dominating surface and solution effects on the fibrillogenic behavior and amyloid assembly of the C-36 peptide, a circulating bioactive peptide from the α