EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Dioscin induces prostate cancer cell apoptosis through activation of estrogen receptor-β.
PMID 28796245
Abstract
Recent researches have shown that estrogen receptor-β (ERβ) activator may be a potent anticancer agent for prostate cancer (PCa), and our previous study also indicated that dioscin can upregulate the expression of ERβ in MC3T3-E1 cell. In the present work, the activity and mechanism of dioscin, a natural product, against PCa were investigated. The results showed that dioscin markedly inhibited cell viability, colony formation, motility and induced apoptosis in PC3 cells. Moreover, dioscin disrupted the formation of PC3 cell-derived mammospheres and reduced aldehyde dehydrogenase (ALDH) level and the CD133