EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Functionalized polymer monoliths with carbamylated amylose for the enantioselective reversed phase nano-liquid chromatographic separation of a set of racemic pharmaceuticals.
PMID 28797665
Abstract
Here we report the first encapsulation of three carbamylated amylose namely R-, S- and R/S-amylose 2,3(3,5-dimethylphenylcarbamate)-6-ethylphenylcarbamate in organic polymer monolith in situ capillary columns. The columns were investigated for the enantioselective nano-liquid chromatographic separation of a set of racemic pharmaceuticals, namely, α- and β-blockers, anti-inflammatory drugs, antifungal drugs, norepinephrine-dopamine reuptake inhibitors, catecholamines, sedative hypnotics, antihistaminics, anticancer drugs, and antiarrhythmic drugs. Baseline separation was achieved for several drugs under reversed phase chromatographic conditions and only few drugs were separated under normal phase conditions. The developed columns provide more economical analysis under environmentally benign conditions.
Related Materials