EMAIL THIS PAGE TO A FRIEND
PMID 28798736
Abstract
Shiga toxin producing
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04058
1-Propanesulfonic acid, technical, ≥99.0% (T)
C3H8O3S