EMAIL THIS PAGE TO A FRIEND
Journal of biomaterials science. Polymer edition
Preparation and characterization of protein-resistant hydrogels for soft contact lens applications via radical copolymerization involving a zwitterionic sulfobetaine comonomer.
PMID 28799461
Abstract
We aimed to introduce hydrophilic sulfobetaine-type zwitterionic groups to macromolecular chains of copolymers to construct novel copolymer hydrogels with anti-protein-fouling performance that could be used as soft contact lens (SCL) materials. Using hydroxyethyl methacrylate (HEMA), N-vinyl pyrrolidinone (NVP) and sulfobetaine methacrylate (SBMA) as comonomers, several copolymer hydrogels with different SBMA contents, poly(HEMA-NVP-SBMA), are synthesized via radical copolymerization in an aqueous phase. Surface chemistry, structural morphologies, water contact angle (WCA), equilibrium water content (EWC), visible light transmittance and tensile mechanical properties are investigated. The prepared hydrogels exhibit a closed-type porous structure. With increasing SBMA content in the comonomer mixture, the prepared copolymer hydrogel pore size gradually increases up to the micron level, WCA tends to decrease, EWC tends to increase, and visible light transmittance slightly increases, but their tensile mechanical properties decline. The amounts of protein Lyz and BSA adsorbed on the copolymer hydrogels and on commercially available EASY DAY
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3409
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
C6H9NO