EMAIL THIS PAGE TO A FRIEND
PMID 28799761
Abstract
Legacy perfluoroalkyl substances (PFASs) are known environmental pollutants with serious adverse health effects. Perfluoroethercarboxylic acids (PFECAs), emerging PFASs now being substituted for legacy PFASs, have recently been detected in the environment. Cyclodextrins (CDs) have been proposed as agents for the remediation of problematic pollutants, including legacy PFASs. The current study uses
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372072
Sodium deuteroxide solution, 40 wt. % in D2O, 99.5 atom % D
DNaO