EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and anticancer properties of ruthenium (II) complexes as potent apoptosis inducers through mitochondrial disruption.
PMID 28800456
Abstract
A new ligand MHPIP (MHPIP = 2-(1-methyl-1H-pyrazol-4-yl)-1H-imidazo[4,5-f][1,10]phenanthroline) and its three ruthenium (II) complexes [Ru(N-N)