PloS one
Extracting information from free-text electronic patient records to identify practice-based evidence of the performance of coronary stents.
PMID 28800599
Abstract
Percutaneous coronary intervention (PCI) using drug-eluting stents (DES) is an indispensable treatment for coronary artery disease. However, to evaluate the performance of various types of stents for PCI, numerous resources are required. We extracted clinical information from free-text records and, using practice-based evidence, compared the efficacy of various DES. We developed a text mining tool based on regular expression and applied it to PCI reports stored in the electronic health records (EHRs) of Ajou University Hospital from 2010-2014. The PCI data were extracted from EHRs with a sensitivity of 0.996, a specificity of 1.000, and an F-measure of 0.995 when compared with a sample of 200 reports. Using these data, we compared the performance of stents by Kaplan-Meier analysis and the Cox hazard proportional regression. In the self-validation analysis comparing the first-generation to the second-generation DES, the second-generation DES was superior to the first-generation DES (hazard ratio [HR]: 0.423, 95% confidence interval [CI]: 0.284-0.630) in terms of target vessel revascularization (TVR), showing similar findings to the established results of previous studies. Among the second-generation DES, the biodegradable-polymer DES tended to be superior, with a risk of TVR (HR: 0.568, 95% CI: 0.281-1.147) falling below than that for the durable-polymer DES approximately 1 year after the index procedure. The Endeavor stent had the highest TVR risk among the newer generation DES (HR: 2.576, 95% CI: 1.273-5.210). In this study, we demonstrated how to construct a PCI data warehouse of PCI-related parameters obtained from free-text electronic records with high accuracy for use in the post surveillance of coronary stents in a time- and cost effective manner. Post surveillance of the practice based evidence in the PCI data warehouse indicated that the biodegradable-polymer DES might have a lower risk of TVR than the durable-polymer DES.