EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Muscle contraction induced arterial shear stress increases endothelial nitric oxide synthase phosphorylation in humans.
PMID 28801524
Abstract
We determined if local increases in brachial artery shear during repetitive muscle contractions induce changes in protein expression of endothelial nitric oxide synthase (eNOS) and/or phosphorylated (p-)eNOS at Ser