EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
A species dependent response to the pro-epileptic drug pentylentetrazole in birds.
PMID 28802899
Abstract
Epilepsy is common disorder that affects over 50 million people worldwide. Birds remain a promising yet largely under-explored model of epilepsy. This study reports the comparison of the response of two species of birds, Australian Parrots (APs) and Sparrows (SPs), to a pro-epileptic drug, Pentylenetetrazole (PTZ). PTZ injections caused myoclonic jerks (MCJs) and tonic clonic seizures (TCSs) in both species. The frequency of MCJs in APs was greater at the dose of 75mg/kg compared to both 50mg/kg and 25mg/kg while it was not significantly different in SPs. The comparison of APs and SPs showed that the frequency of MCJs was greater in APs compared to SPs at 25mg/kg and 75mg/kg while its latency was reduced at 25mg/kg and 50mg/kg. Interestingly SPs had a reduced latency of TCSs compared to APs at 75mg/kg. Glutamatergic and Gabaergic cell count was conducted to determine an association with the epileptic response to PTZ. The Glutamatergic cell counts for SPs was significantly greater than APs and conversely the Gabaergic cell counts in APs was higher compared to SPs. The reason for this difference in findings needs to be further investigated. This study shows that birds, and APs and SPs in particular, are a valid, interesting and under-explored model of epilepsy that should be further explored in order to understand the mysteries of epilepsy.