EMAIL THIS PAGE TO A FRIEND
Life sciences
Role of canonical transient receptor potential channel-3 in acetylcholine-induced mouse airway smooth muscle cell proliferation.
PMID 28802903
Abstract
Canonical transient receptor potential channel-3 (TRPC3)-encoded Ca Primary mouse ASMCs were cultured with or without ACh treatment, then cell viability, TRPC3 expression, NSCC currents and [Ca TRPC3 blocker Gd Our data suggested ACh could induce ASMC proliferation, and TRPC3 may be involved in ACh-induced ASMC proliferation that occurs with airway remodeling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU031761 MISSION® esiRNA, esiRNA human TRPC3 (esiRNA1)