EMAIL THIS PAGE TO A FRIEND
Biochemistry
Identification of the Dimer Exchange Interface of the Bacterial DNA Damage Response Protein UmuD.
PMID 28806503
Abstract
The Escherichia coli SOS response, an induced DNA damage response pathway, confers survival on bacterial cells by providing accurate repair mechanisms as well as the potentially mutagenic pathway translesion synthesis (TLS). The umuD gene products are upregulated after DNA damage and play roles in both nonmutagenic and mutagenic aspects of the SOS response. Full-length UmuD is expressed as a homodimer of 139-amino-acid subunits, which eventually cleaves its N-terminal 24 amino acids to form UmuD'. The cleavage product UmuD' and UmuC form the Y-family polymerase DNA Pol V (UmuD'