EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
A simple high performance liquid chromatography-mass spectrometry method for Therapeutic Drug Monitoring of isavuconazole and four other antifungal drugs in human plasma samples.
PMID 28806568
Abstract
Triazoles chanced the prevention and treatment of invasive fungal infections, but their pharmacokinetic properties are still unclear. In particular, isavuconazole (ISC) is a new broad-spectrum antifungal triazole approved in 2015 as first-line treatment for intravenous and oral use against invasive aspergillosis and for mucormycosis. Nowadays, the optimal management of the treatments with triazoles requires the use of Therapeutic Drug Monitoring (TDM), in order to prevent sub-therapeutic or toxic concentrations. In turn, the routine use of TDM requires reliable quantification methods The aim of this work was the development and full validation of a HPLC-mass spectrometry assay for the simultaneous quantification of fluconazole, itraconazole, isavuconazole, posaconazole and voriconazole in human samples. Both standards and quality controls were prepared in human plasma. After the addition of internal standard (6,7-dimethyl-2,3-di(2-pyridyl)quinaxoline for voriconazole, posaconazole and itraconazole; stable isotope labeled compounds for fluconazole and isavuconazole), protein precipitation with acetonitrile and dilution with water were performed. Chromatographic separation was performed on Atlantis