EMAIL THIS PAGE TO A FRIEND
eLife
RIPK1-RIPK3-MLKL-dependent necrosis promotes the aging of mouse male reproductive system.
PMID 28807105
Abstract
A pair of kinases, RIPK1 and RIPK3, as well as the RIPK3 substrate MLKL cause a form of programmed necrotic cell death in mammals termed necroptosis. We report here that male reproductive organs of both
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2100 RIPA-56, ≥98% (HPLC)
C13H19NO2