EMAIL THIS PAGE TO A FRIEND
Cell calcium
Regulation of L-type Ca
PMID 28807144
Abstract
cGMP is a second messenger widely used in the nervous system and other tissues. One of the major effectors for cGMP is the serine/threonine protein kinase, cGMP-dependent protein kinase (PKG), which catalyzes the phosphorylation of a variety of proteins including ion channels. Previously, it has been shown that the cGMP-PKG signaling pathway inhibits Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1381
8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt, ≥98% (HPLC), powder
C10H10BrN5NaO7P