EMAIL THIS PAGE TO A FRIEND
The Plant cell
Mutations in eIF5B Confer Thermosensitive and Pleiotropic Phenotypes via Translation Defects in
PMID 28808135
Abstract
The conserved eukaryotic translation initiation factor 5B, eIF5B, is a GTPase that acts late in translation initiation. We found that an