EMAIL THIS PAGE TO A FRIEND
Biology open
Differential expression of sirtuin 2 and adipocyte maturation restriction: an adaptation process during hypoxia in fish.
PMID 28808139
Abstract
Sirtuins have received widespread attention due to their diverse physiological role in metabolism. Among sirtuins, SIRT2 is more abundant in adipocytes and exerts effects on adipocyte differentiation, a process which involves conversion of preadipocytes to mature adipocytes orchestrated by adipokines and adipogenic transcription factors. Grey mullet (