EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Physicochemical and adsorptive characteristics of activated carbons from waste polyester textiles utilizing MgO template method.
PMID 28808853
Abstract
Activated carbons with high specific surface areas were produced, utilizing waste polyester textiles as carbon precursor by magnesium oxide (MgO) template method. Magnesium chloride (MgCl
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS000001
Magnesium citrate, AldrichCPR
C6H6MgO7