ACS applied materials & interfaces
Nano-in-Nano Approach for Enzyme Immobilization Based on Block Copolymers.
PMID 28809474
We set up a facile approach for fabrication of supports with tailored nanoporosity for immobilization of enzymes. To this aim block copolymers (BCPs) self-assembly has been used to prepare nanostructured thin films with well-defined architecture containing pores of tailorable size delimited by walls with tailorable degree of hydrophilicity. In particular, we employed a mixture of polystyrene-block-poly(l-lactide) (PS-PLLA) and polystyrene-block-poly(ethylene oxide) (PS-PEO) diblock copolymers to generate thin films with a lamellar morphology consisting of PS lamellar domains alternating with mixed PEO/PLLA blocks lamellar domains. Selective basic hydrolysis of the PLLA blocks generates thin films, patterned with nanometric channels containing hydrophilic PEO chains pending from PS walls. The shape and size of the channels and the degree of hydrophilicity of the pores depend on the relative length of the blocks, the molecular mass of the BCPs, and the composition of the mixture. The strength of our approach is demonstrated in the case of physical adsorption of the hemoprotein peroxidase from horseradish (HRP) using 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) with H