EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Beneficial effects of dried pomegranate juice concentrated powder on ultraviolet B-induced skin photoaging in hairless mice.
PMID 28810554
Abstract
The present study investigated the anti-aging effects of pomegranate juice concentrated powder (PCP) in hairless mice following 15 weeks of UVB irradiation (three times a week; 0.18 J/cm