EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Combination of alendronate and genistein synergistically suppresses osteoclastic differentiation of RAW267.4 cells
PMID 28810648
Abstract
Bone is a dynamic tissue that undergoes constant remodeling, with removal by osteoclastic bone resorption and replacement via osteoblastic bone formation and mineralization. Deterioration of bone mass with aging leads to osteoporosis. Bisphosphonates are potent inhibitors of osteoclastic bone resorption. Genistein, an isoflavone, exerts a bone anabolic effect by suppressing osteoclastic bone resorption and stimulating osteoblastic bone formation. The present study was undertaken to investigate the anabolic effects of a combination of alendronate and genistein on osteoclastic differentiation. Preosteoclastic RAW267.4 cells were cultured with alendronate (0.1-100 µM) and/or genistein (0.1-100 µM)