EMAIL THIS PAGE TO A FRIEND
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques
Clofibrate Attenuates ROS Production by Lipid Overload in Cultured Rat Hepatoma Cells.
PMID 28810950
Abstract
To investigate the effect of clofibrate on inducing liver fatty acid binding protein (FABP1) following a high-fat load in a hepatocyte cell culture model. Rat hepatoma cells (CRL-1548) were treated with a fatty acid (FA) mixture consisting of oleate:palmitate (2:1) in the presence of 3% albumin. Cells were treated with 0, 0.5, 1, 2, or 3 mM FA for 24 and 48 hr, or further treated with 500 µM clofibrate (CLO) to induce FABP1 levels. Cytotoxicity was determined using the WST-1 assay. Intracellular lipid droplets were quantitated following staining with Nile Red. Dichlorofluorescein (DCF) was used to assess the extent of intracellular reactive oxygen species (ROS). Cell viability decreased (p < 0.01) with an increase in lipid concentration. Intracellular lipid droplets accumulated significantly (p < 0.001) with an increase in long-chain fatty acid load, which was associated with a statistical increase (p < 0.05) in ROS levels. Early clofibrate treatment showed significant increases in intracellular FABP1 levels with significant decreases in ROS levels (p < 0.05). Silencing FABP1 expression using siRNA revealed that FABP1 was the main contributor for the observed intracellular ROS clearance. Characteristic cellular damage resulted from released ROS following a high fat load to hepatoma cells. The damage was attenuated through early treatment with clofibrate, which may act as a hepatoprotectant by inducing FABP1 expression and in this manner, suppress intracellular ROS levels. This article is open to POST-PUBLICATION REVIEW. Registered readers (see "For Readers") may comment by clicking on ABSTRACT on the issue's contents page.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU122491 MISSION® esiRNA, esiRNA human FABP1 (esiRNA1)