EMAIL THIS PAGE TO A FRIEND
Clinical rheumatology
Increased IL-6 expression on THP-1 by IL-34 stimulation up-regulated rheumatoid arthritis Th17 cells.
PMID 28812210
Abstract
IL-34 is a pleiotropic cytokine, which is a key regulator of monocytes/macrophages and might participate in the pathogenesis of RA. In this study, we aimed to explore the effect of IL-34 on the monocyte-like cell line THP-1 and the quantitative variation of Th17 cells in THP-1 and RA CD4