EMAIL THIS PAGE TO A FRIEND
Scientific reports
Differential oxidation of protein-tyrosine phosphatases during zebrafish caudal fin regeneration.
PMID 28814789
Abstract
Zebrafish have the capacity to regenerate lost tissues and organs. Amputation of the caudal fin results in a rapid, transient increase in H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1885
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder
C16H20N2O8