EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Hepatoprotective and antioxidant effects of single clove garlic against CCl
PMID 28818066
Abstract
The increase in demand and consumption of single clove garlic or 'Solo garlic' (Allium sativum) has resulted in an increase in research on its therapeutic properties. The present study aims to evaluate the antioxidant activities, oxidant-scavenging efficiency and preventive effects of SCG (single clove garlic) and MCG (multi clove garlic) on CCl For this purpose, rabbits were orally administered with 3 ml of CCl SCG extracts exhibited higher antioxidant capacity than the MCG extract. Scavenging ability of SCG showed significant (p < 0.05) elevation against 2,2-diphenyl-1-picrylhydrazyl (DPPH) and superoxide radicals in comparison to MCG. In addition, total phenolic content of SCG was significantly elevated (p < 0.001), thereby suggesting that the composition of garlic storage constituents varies with the number of cloves present. CCl The present study indicates that SCG possesses more protective ability than MCG against CCl